Imprimer

R-(+)-Baclofen hydrochloride ≥98% (par HPLC)

Fournisseur: Enzo Life Sciences

ALEXALX550074M050EA 0 CHF
ALEXALX550074M050
R-(+)-Baclofen hydrochloride ≥98% (par HPLC)
R-(+)-Baclofen hydrochloride

Selective GABAB receptor agonist.


Skeletal muscle relaxant. The more active enantiomer of (±)-baclofen.


Soluble in water, dilute aqueous base or DMSO.

Formule: C₁₀H₁₂ClNO₂·HCl
MW: 250,2 g/mol
température de stockage: Réfrigérateur
Numéro CAS: 63701-55-3
https://ch.vwr.com/stibo/web/std.lang.all/08/00/45390800.jpg

Order Now

Résultats des tests de spécification

Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR