Imprimer

8-Chloroadenosine-3',5'-cyclic-monophosphate ≥98% (par HPLC)

ALEXALX480029M001EA 67 CHF
ALEXALX480029M001
8-Chloroadenosine-3',5'-cyclic-monophosphate ≥98% (par HPLC)
8-Chloroadenosine-3',5'-cyclic-monophosphate

PKA activator.


cAMP analog with high selectivity for cAMP-dependent protein kinase type I. Induces growth inhibition and differentiation in cultured cancer cell lines.


At least 200 mM soluble in water, aqueous buffers, methanol, DMSO and dimethylformamide; sparingly soluble in 100% ethanol (13 mM). May produce hazy solutions.

Formule: C₁₀H₁₀ClN₅NaO₆P
MW: 385,633 g/mol
température de stockage: Congélateur
Numéro CAS: 41941-56-4

Order Now

Résultats des tests de spécification

Purity (HPLC) ≥98%
Identity Identity determined by ³¹P-NMR, ¹H-NMR, ESI-MS and UV

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR