Imprimer

8-Bromoadenosine-3',5'-cyclic monophosphothioate, Sp-isomer sodium salt ≥98% (par HPLC)

Fournisseur: Enzo Life Sciences

ALEXALX480025M005EA 851 CHF
ALEXALX480025M005 ALEXALX480025M001
8-Bromoadenosine-3',5'-cyclic monophosphothioate, Sp-isomer sodium salt ≥98% (par HPLC)
8-Bromoadenosine-3',5'-cyclic monophosphothioate, Sp-isomer sodium salt

PKA activator.


Metabolically resistant activator of cAMP-dependent protein kinase (PKA) (cAMP agonist).


No metabolic side effects as observed with dibutyryl-cAMP (N⁶,O²'-dibutyryladenosine-3',5'-cyclic monophosphate, or 8-Br-cAMP (8-bromoadenosine-3',5'-cyclic monophosphate. Significantly more lipophilic and membrane permeant as compared to Sp-cAMPS (adenosine-3',5'-cyclic monophosphothioate, Sp isomer).


Soluble in water or aqueous buffer.

Formule: C₁₀H₁₀BrN₅O₅PS·Na
MW: 446,2 g/mol
température de stockage: Congélateur
Numéro CAS: 127634-20-2
https://ch.vwr.com/stibo/web/std.lang.all/07/66/45390766.jpg

Order Now

Résultats des tests de spécification

Purity (HPLC) ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR