Ochratoxin A ≥98% (par HPLC, TLC)
Fournisseur: Enzo Life Sciences
|
Danger
|
Mycotoxin. Nephrotoxic.
Mycotoxin. Natural contaminant of mouldy food and feed. It has a number of toxic effects, the most prominent being nephrotoxicity. Immunosuppressive, genotoxic, teratogenic and carcinogenic. Stimulates lipid peroxidation. Induces oxidative DNA lesions coupled with direct DNA adducts via quinone formation.
Isolated from Aspergillus ochraceus.
Soluble in methanol (10 mg/ml).
Formule:
C₂₀H₁₈ClNO₆ MW: 403,8 g/mol température de stockage: Réfrigérateur |
MDL Number:
MFCD00078079 Numéro CAS: 303-47-9 ONU: 2811 ADR: 6.1,II |
Résultats des tests de spécification
Purity | ≥98% (HPLC, TLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...