Imprimer
20-Hydroxyleukotriene B4 ≥99% (par HPLC)
Fournisseur: Enzo Life Sciences
Danger
ALEXBMLHLB004-0050EA 0 CHF
ALEXBMLHLB004-0050
20-Hydroxyleukotriene B4 ≥99% (par HPLC)
20-Hydroxyleukotriene B4
Primary LTB4 metabolite produced by ω oxidation in PMNL retaining some chemotactic activity. ω oxidation is blocked by 17-ODYA.
Formule: C₂₀H₃₂O₅
température de stockage: Congélateur ultra-basse température (–80°C)
Numéro CAS: 79516-82-8
ONU: 1170
ADR: 3,II
https://ch.vwr.com/stibo/web/std.lang.all/12/85/45401285.jpg
Order Now
Résultats des tests de spécification
Source Synthetic
Concentration 50 µg/ml
Formulation Solution in ethanol
Purity ≥99% (HPLC)