Imprimer

Mito-TEMPO ≥95% (par CCM)

Fournisseur: Enzo Life Sciences
Attention

ALEXALX430150-M005EA 0 CHF
ALEXALX430150-M005
Mito-TEMPO ≥95% (par CCM)
Mito-TEMPO

Mitochondria-targeted antioxidant with superoxide and alkyl radical scavenging properties, which may be used for both in vivo and in vitro experiments.


Soluble in water, DMSO, 100% ethanol or methanol.

Formule: C₂₉H₃₅N₂O₂ClP·H₂O
MW: 528,1 g/mol
température de stockage: Congélateur
Numéro CAS: 1569257-94-8
https://ch.vwr.com/stibo/web/std.lang.all/06/99/45390699.jpg

Order Now

Résultats des tests de spécification

Purity (TLC) ≥95%
Identity Determined by MS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR