Imprimer

(+)-Himbacine ≥98%, en poudre

Fournisseur: Enzo Life Sciences

ALEXALX550061M001EA 0 CHF
ALEXALX550061M001 ALEXALX550061M005 ALEXALX550061M025
(+)-Himbacine ≥98%, en poudre
(+)-Himbacine

Muscarinic acetylcholine receptor antagonist.


Potent muscarinic antagonist that displays selectivity for the M2 or M4 receptors.


Hygroscopic and soluble in methanol, 100% ethanol or methylene chloride; insoluble in water.

Formule: C₂₂H₃₅NO₂
température de stockage: Température ambiante
Numéro CAS: 6879-74-9
https://ch.vwr.com/stibo/web/std.lang.all/07/98/45390798.jpg

Order Now

Résultats des tests de spécification

Purity ≥98%
Source Isolated from the Australian pine galbulimima baccata
Appearance White to off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR