Imprimer

8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt ≥99% (par HPLC)

Fournisseur: Enzo Life Sciences

ALEXALX480088M005EA 0 CHF
ALEXALX480088M005 ALEXALX480088M001
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt ≥99% (par HPLC)
8-(4-Chlorophenylthio)guanosine-3',5'-cyclic monophosphate sodium salt

Selective activator of cGMP-dependent protein kinase (PKG) Iα, Iβ, and type II as well as of cGMP-gated ion channels.


Higher activation potential as compared to cGMP; excellent cell membrane permeability and phosphodiesterase stability. Preferred alternative to unsuitable analogs such as dibutyryl-cGMP.


Soluble in water.

Formule: C₁₆H₁₄ClN₅O₇PS·Na
MW: 509,8 g/mol
température de stockage: Congélateur
Numéro CAS: 51239-26-0
https://ch.vwr.com/stibo/web/std.lang.all/07/77/45390777.jpg

Order Now

Résultats des tests de spécification

Purity (HPLC) ≥99%
Identity Determined by ESI-MS and UV/VIS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR