Imprimer

Trimethylammonio-PTIO methylsulphate ≥80%

Fournisseur: Enzo Life Sciences

ALEXALX430085M050EA 0 CHF
ALEXALX430085M050
Trimethylammonio-PTIO methylsulphate ≥80%
Trimethylammonio-PTIO methylsulphate

Spin trapping reagent.


Nitric oxide (NO) spin trap that is non-cell permeable, allowing study of extracellular NO release.


Soluble in water, 100% ethanol or DMSO.

https://ch.vwr.com/stibo/web/std.lang.all/06/68/45390668.jpg

Order Now

Résultats des tests de spécification

Purity (assay) ≥80%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR