Imprimer

Calcimycin (mixed Mg/CA salt) ≥98% (par CCM)

Fournisseur: Enzo Life Sciences
Danger

ALEXALX450002M010EA 0 CHF
ALEXALX450002M010 ALEXALX450002M005
Calcimycin (mixed Mg/CA salt) ≥98% (par CCM)
Calcimycin (mixed Mg/CA salt)

Salt form of mixed calcium-magnesium. Used in cell activation experiments when calcium dose-response data are not required.


Soluble in methylene chloride, chloroform, acetone, ethyl acetate, or diethyl ketone; also soluble in DMSO, 100% ethanol, and ethanol. Solubilise in DMSO or ethanol prior to preparing aqueous solutions of very low concentrations.

Formule: (C₂₉H₃₆N₃O₆)₂·Mg(C₂₉H₃₆N₃O₆)₂·Ca
température de stockage: Réfrigérateur
Numéro CAS: 52665-69-7

Order Now

Résultats des tests de spécification

Purity (TLC) ≥98%
Source Isolated from Streptomyces chartreusensis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR