Sapintoxin D (high purity) ≥98% (par HPLC)
Fournisseur: Enzo Life Sciences
|
Danger
|
ALEXALX445047M001EA
0
CHF
ALEXALX445047M001
ALEXALX445047M005
Sapintoxin D (high purity) ≥98% (par HPLC)
Sapintoxin D (high purity)
Sapintoxin D act as a PKC ligand. Fluorescent, highly potent phorbol ester.
Useful for membrane studies.
Soluble in methanol, DMSO or chloroform.
Formule:
C₃₀H₃₇NO₈ température de stockage: Congélateur |
Numéro CAS:
80998-07-8 ONU: 3462 ADR: 6.1,I |
Résultats des tests de spécification
Purity (HPLC) | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...