Imprimer

Leukotriene A4 methyl ester ≥98% (par HPLC)

Fournisseur: Enzo Life Sciences
Danger

ALEXBMLLPA004-1000EA 0 CHF
ALEXBMLLPA004-1000 ALEXBMLLPA004-0050
Leukotriene A4 methyl ester ≥98% (par HPLC)
Leukotriene A4 methyl ester

Biosynthesis in rabbit reticulocytes and human leukocytes.

Formule: C₂₁H₃₂O₃
température de stockage: Congélateur ultra-basse température (–80°C)
Numéro CAS: 73466-12-3
ONU: 3082
ADR: 9,III
https://ch.vwr.com/stibo/web/std.lang.all/13/53/45401353.jpg

Order Now

Résultats des tests de spécification

Purity ≥98% (HPLC)
Concentration 50 µg/ml
Formulation Oil dissolved in 2% triethylamine/hexane

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR