NOR-5 ≥98% (par HPLC)
Fournisseur: Enzo Life Sciences
|
Attention
|
ALEXALX430125M010EA
0
CHF
ALEXALX430125M010
ALEXALX430125M005
NOR-5 ≥98% (par HPLC)
NOR-5
Nitric oxide (NO) donor.
The half-life time of 0.5 mM NOR-5 is 20 hours at 37 °C in 0.1 M PBS, pH 7.4.
Soluble in DMSO (10 mg/ml).
Résultats des tests de spécification
Purity (HPLC) | ≥98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...