Imprimer
ALEXALX500002
ALEXALX500002L100EA 1107 CHF
ALEXALX500002L100 ALEXALX500002L025 ALEXALX500002L005
ALEXALX500002
Non-ionic detergent.
n-Octyloligooxyethylene is used as a detergent for solubilisation, purification and crystallisation of membrane proteins.