Lysophosphatidic acid ≥98% (par CCM)
Fournisseur: Enzo Life Sciences
LPA receptor ligand
Lysophosphatidic acid (LPA) is a multifunctional intercellular phospholipid messenger.
LPA stimulates the growth of a variety of cells including fibroblasts, vascular smooth muscle cells, endothelial cells and keratinocytes. It acts as a proliferative and anti-apoptotic factor. Ligand for LPA1 (EDG-2), LPA2 (EDG-4) and LPA3 (EDG-7) receptors. It also inhibits differentiation of neural stem cells into neurons.
Soluble in water (5 mg/ml with sonication) or DMSO (0.5 mg/ml warm).
Formule:
C₂₁H₄₀O₇P·Na MW: 458,5 g/mol température de stockage: Congélateur |
MDL Number:
MFCD00133427 Numéro CAS: 325465-93-8 |
Résultats des tests de spécification
Purity | ≥98% (TLC) |
Appearance | Waxy solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...