Imprimer

CPH hydrochloride ≥99% (EPR), solide blanc à blanc cassé

Fournisseur: Enzo Life Sciences

ALEXALX430078M010EA 0 CHF
ALEXALX430078M010 ALEXALX430078M250 ALEXALX430078M050
CPH hydrochloride ≥99% (EPR), solide blanc à blanc cassé
CPH hydrochloride

Spin trapping reagent.


  • Effective, cell permeable and non-toxic spin probe for the detection of superoxide radical and peroxynitrite, both in vitro and in vivo.


Resistant to reduction by vitamin C and thiols.


Soluble in DMSO, 100% ethanol, methanol or water.

Formule: C₉H₁₇NO₃·HCl
MW: 223,7 g/mol
température de stockage: Congélateur
Numéro CAS: 92416-28-9
https://ch.vwr.com/stibo/web/std.lang.all/06/62/45390662.jpg

Order Now

Résultats des tests de spécification

Purity (EPR) ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR