Imprimer

DL-Tryptophane

Fournisseur: Molekula

MOLEM52045958EA 0 CHF
MOLEM52045958
DL-Tryptophane
DL-Tryptophane
Formule: C₁₁H₁₂N₂O₂
MW: 204,23 g/mol
Melting Pt: 289…290 °C
Densité: 1,349 g/cm³ (20 °C)
MDL Number: MFCD00064339
Numéro CAS: 54-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR