Imprimer

D-erythro-Dihydrosphingosine ≥98%

Fournisseur: Enzo Life Sciences

ALEXBML-SL125-0050EA 0 CHF
ALEXBML-SL125-0050 ALEXBML-SL125-0010
D-erythro-Dihydrosphingosine ≥98%
D-erythro-Dihydrosphingosine

Sphingosine precursor


Biosynthetic precursor of sphingosine. Inhibitor of protein kinase C (PKC). Blocks phospholipases A2 (PLA2) and the D-sphingosine precursor.

Formule: C₁₈H₃₉NO₂
MDL Number: MFCD07371371
Numéro CAS: 764-22-7
https://ch.vwr.com/stibo/web/std.lang.all/15/14/45401514.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR