Imprimer

Tetrodotoxin ≥98% (par CCM)

Fournisseur: Enzo Life Sciences

ALEXBML-NA120-0001EA 0 CHF
ALEXBML-NA120-0001
Tetrodotoxin ≥98% (par CCM)
Tetrodotoxin

Na+ channel blocker


Tetrodotoxin is a non-peptide toxin isolated from ovaries and liver of chinase puffer fish. Neurotoxin. Potent and reversible inhibitor of the voltage dependent Na+ channels in nerve membranes.

Formule: C₁₁H₁₇N₃O₈
MW: 319,27 g/mol
MDL Number: MFCD00213719
Numéro CAS: 4368-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR