Morin ≥85%
Fournisseur: Enzo Life Sciences
ALEXALX385016G001EA
0
CHF
ALEXALX385016G001
Morin ≥85%
Morin
Cell cycle inhibitor
Antioxidant flavonoid. Shows anti-proliferative and antitumor properties. Induces apoptosis. Induces cell cycle arrest at the G2/M phase. Anti-inflammatory compound. Suppresses NF-κB activation. Induces lipid peroxidation and DNA strand breaks.
Formule:
C₁₅H₁₀O₇ MW: 302,24 g/mol |
MDL Number:
MFCD00217054 Numéro CAS: 480-16-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...