Verruculogen ≥96% (par HPLC, TLC)
Fournisseur: Enzo Life Sciences
ALEXALX380056M001EA
0
CHF
ALEXALX380056M001
ALEXALX380056M005
Verruculogen ≥96% (par HPLC, TLC)
Verruculogen
Cell cycle inhibitor
Neurotoxin that acts as an inhibitor of the M phase of the mammalian cell cycle. Inhibits Ca2+-activated K+ channels. Decreases GABA levels in CNS.
Isolated from Penicillium verruculosum.
Formule:
C₂₇H₃₃N₃O₇ MW: 511,6 g/mol |
Numéro CAS:
12771-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...