Imprimer

DL-sulphoraphane (synthétique) ≥98% (par UPLC)

Fournisseur: Enzo Life Sciences

ALEXALX350232M025EA 0 CHF
ALEXALX350232M025
DL-sulphoraphane (synthétique) ≥98% (par UPLC)
DL-sulphoraphane

R,S-Sulforaphane is a synthetic version of the naturally occurring compound, R-sulforaphane, from Broccoli. It is a racemic mixture of both the R and S isomers.
Potent, selective inducer of phase II detoxification enzymes. Inhibits chemically induced mammary tumor formation in rats. Inhibits LPS-induced HMGB1 (high mobility group box 1) secretion.


  • Carcinogenesis inhibitor


Slightly yellowish liquid.


Soluble in methanol or DMSO.

Formule: C₆H₁₁NOS₂
MW: 177,3 g/mol
température de stockage: Congélateur
MDL Number: MFCD00198068
Numéro CAS: 4478-93-7
https://ch.vwr.com/stibo/web/std.lang.all/77/14/45347714.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR