Imprimer

Fluvastatin sodium salt ≥98% (par HPLC)

Fournisseur: Enzo Life Sciences

ALEXALX270466M010EA 0 CHF
ALEXALX270466M010 ALEXALX270466M050
Fluvastatin sodium salt ≥98% (par HPLC)
Fluvastatin sodium salt

Anticholesterol agent. HMG-CoA inhibitor.


Anticholesterol agent. HMG-CoA reductase inhibitor and antioxidant.


Synthetic.

Formule: C₂₄H₂₅FNNaO₄
MW: 433,46 g/mol
MDL Number: MFCD00929076
Numéro CAS: 93957-55-2
https://ch.vwr.com/stibo/web/std.lang.all/75/87/45347587.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR