Imprimer

SB202190 hydrochloride ≥97%

Fournisseur: Enzo Life Sciences

ALEXALX270268M001EA 0 CHF
ALEXALX270268M001 ALEXALX270268M005
SB202190 hydrochloride ≥97%
SB202190 hydrochloride

p38 inhibitor


A water soluble form of the potent p38 (SAPK2a) inhibitor SB202190 (Prod. No. BML-EI294).

Formule: C₂₀H₁₄FN₃O·HCl
MW: 367,9 g/mol
Numéro CAS: 350228-36-3
https://ch.vwr.com/stibo/web/std.lang.all/75/05/45347505.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR