Imprimer

Ophiobolin A

Fournisseur: Enzo Life Sciences

ALEXALX270109M001EA 0 CHF
ALEXALX270109M001
Ophiobolin A
Ophiobolin A

Calmodulin antagonist. PDE inhibitor.


Cell permeable antagonist of calmodulin. Inhibits Ca2+/calmodulin-dependent phosphodiesterase. Inhibits P-glycoprotein-mediated transport. Exhibits antibacterial, antitumor and nematocidal activities. Phytotoxic.


Isolated from Bipolaris leersia MST-FP107.

Formule: C₂₅H₃₆O₄
MW: 400,6 g/mol
MDL Number: MFCD00058400
Numéro CAS: 4611-05-6
https://ch.vwr.com/stibo/web/std.lang.all/74/79/45347479.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR