Imprimer

Bisindoylmaleimide X hydrochloride ≥98%

Fournisseur: Enzo Life Sciences

ALEXALX270057M001EA 0 CHF
ALEXALX270057M001 ALEXALX270057M005
Bisindoylmaleimide X hydrochloride ≥98%
Bisindoylmaleimide X hydrochloride

PKC inhibitor


Selective inhibitor of protein kinase C (PKC).

Formule: C₂₆H₂₄N₄O₂·HCl
MW: 461 g/mol
MDL Number: MFCD04972010
Numéro CAS: 145317-11-9
https://ch.vwr.com/stibo/web/std.lang.all/74/68/45347468.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR