Imprimer

(6S)-Tetrahydrofolic acid ≥95%

Attention

CAYM18263-100EA 856 CHF
CAYM18263-100 CAYM18263-50 CAYM18263-25
(6S)-Tetrahydrofolic acid ≥95%
(6S)-Tetrahydrofolic acid

(6S)-Tetrahydrofolic acid is a diastereomer of tetrahydrofolic acid, a reduced form of folic acid that serves as a cofactor in methyltransferase reactions and is the major one-carbon carrier in one carbon metabolism.
(6S)-Tetrahydrofolic acid is 1,000-fold more active than the (6R) form at promoting the binding of fluorodeoxyuridylate to thymidylate synthase and 600-fold more active as a growth factor of P. cerevisiae.
(6S)-Tetrahydrofolic acid also has a low affinity and high dissociation rate for folate-binding protein.3


  • Stability : ≥ 2 years at -20°C
  • Supplied as: A crystalline solid
  • UV visible: λ max 300 nm

Formule: C₁₉H₂₃N₇O₆
MW: 445,43 g/mol
température de stockage: Congélateur
Numéro CAS: 71963-69-4
EINECS: 205-181-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR