Imprimer

Mometasone furoate

Fournisseur: Thermo Scientific
Danger

ACRO462130010EA 150 CHF
ACRO462130010
Mometasone furoate
Mometasone furoate
Formule: C₂₇H₃₀Cl₂O₆
Melting Pt: 218…220 °C
température de stockage: Réfrigérateur
MDL Number: MFCD00866003
Numéro CAS: 83919-23-7

Order Now

Résultats des tests de spécification

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 216 to 222 °C
Loss on drying ≤0.5 % (105 °C)
HPLC ≥96.0 %
Specific optical rotation +48° to +55° (20 °C, 589 nm) (0.5% in Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR