Imprimer

(1-Formylcyclopropyl)carbamic acid-tert-butyl ester 97%

Fournisseur: Thermo Scientific
Danger

ACRO449950010EA 382 CHF
ACRO449950010
(1-Formylcyclopropyl)carbamic acid-tert-butyl ester 97%
(1-Formylcyclopropyl)carbamic acid-tert-butyl ester
Formule: C₉H₁₅NO₃
MW: 185,22 g/mol
température de stockage: Température ambiante
MDL Number: MFCD10697899
Numéro CAS: 107259-06-3
ONU: 3077
ADR: 9,III

Order Now

Résultats des tests de spécification

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 82 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR