Imprimer

Nichrome solution d'attaque

Fournisseur: Thermo Scientific
Danger

44585.WYEA 154 CHF
44585.WY
Nichrome solution d'attaque
Nichrome

Nichrome Etchant is a high purity system based on ceric ammonium nitrate. It provides clean, precise etching of evaporated nichrome layers for the preparation of thin film circuits. This product can be operated at room temperature or elevated temperature to increase etch rate.


Semiconductor-grade ingredients for ultra-high purity
Etchant filter to 0,2 microns to remove particles

Densité: 1,25 g/cm³ (20 °C)
température de stockage: Température ambiante
MDL Number: MFCD07370976
ONU: 3264
ADR: 8,III

Order Now

Résultats des tests de spécification

Appearance Clear orange
Chemistry base Ceric ammonium nitrate
Specific gravity 1,127 - 1,130
Filtration 0,2 microns
Operating temperature 40 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR