Imprimer

Anti-HEPACAM Rabbit Polyclonal Antibody (Cy3®)

Fournisseur: Bioss
BOSSBS-5840R-CY3EA 1140 CHF
BOSSBS-5840R-CY3
Anti-HEPACAM Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Involved in regulating cell motility and cell-matrix interactions. May inhibit cell growth through suppression of cell proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HEPACAM
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR