Imprimer

Anti-DEFA1 Rabbit Polyclonal Antibody (Cy5.5®)

Fournisseur: Bioss
BOSSBS-1697R-CY5.5EA 1150 CHF
BOSSBS-1697R-CY5.5
Anti-DEFA1 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorps
Defensins are a group of cyclic peptides containing 29-35 amino acids, which comprise approximately 30% of the total protein content of neutrophil azurophilic granules. Various functions have been reported for the defensins, including antibacterial and antifungal activity and chemotactic activity for monocytes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DEFA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR