Imprimer

Abamectin ≥97%, en poudre

Danger

J60039.06EA 425 CHF
J60039.06 J60039.03 J60039.14
Abamectin ≥97%, en poudre
Abamectin

Abamectin interacts with GABA receptors to cause chloride channel activation. Used to control a range of agronomic, fruit, vegetable and ornamental crops. Also it is used for control of fire ants and in veterinary antihelmintic.


Used to control a range of agronomic, fruit, vegetable and ornamental crops. Also it is used for control of fire ants and in veterinary antihelmintic.

Formule: C₄₈H₇₂O₁₄
MW: 873,10 g/mol
Melting Pt: 150…155 °C
température de stockage: Réfrigérateur
MDL Number: MFCD01769550
Numéro CAS: 71751-41-2
ONU: 2811
ADR: 6.1,I

Order Now

Résultats des tests de spécification

Form Powder
Assay (unspecified) 0,2% max
Assay (HPLC) 75% min (Avermectin B1)
Assay (HPLC) 70% min (Avermectin B1a)
Loss on Drying 2,0% max
Solubility Soluble in DMSO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR