Imprimer

Peracetic acid ∼38 - 40%, Supelco®

Fournisseur: Merck
Danger


M-Clarity™ quality level = MQ200

1.07222.1000EA 295 CHF
1.07222.1000
Peracetic acid ∼38 - 40%, Supelco®
Peracetic acid
Formule: C₂H₄O₃
MW: 76,051 g/mol
température de stockage: Température ambiante
MDL Number: MFCD00002128
Numéro CAS: 79-21-0
ONU: 3105
ADR: 5.2 (8),

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR