Imprimer

1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Fournisseur: Thermo Scientific
Attention

ACRO444440010EA 196 CHF
ACRO444440010
1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester
Formule: C₁₁H₁₇BN₂O₃
MW: 236,08 g/mol
Melting Pt: 47…49 °C
température de stockage: Température ambiante
MDL Number: MFCD09027064
Numéro CAS: 1150561-76-4

Order Now

Résultats des tests de spécification

Appearance White to light yellow Solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR