Imprimer

(S)-N-Fmoc-α-methylvaline 98%, 98% ee

Fournisseur: Thermo Scientific
Attention

ACRO441155000EA 495 CHF
ACRO441155000
(S)-N-Fmoc-α-methylvaline 98%, 98% ee
(S)-N-Fmoc-alpha-methylvaline
Formule: C₂₁H₂₃NO₄
MW: 353,418 g/mol
Melting Pt: 37…53 °C
température de stockage: Réfrigérateur
MDL Number: MFCD08752490
Numéro CAS: 169566-81-8
ONU: 3175
ADR: 4.1,II

Order Now

Résultats des tests de spécification

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Enantiomeric excess ≥98.0 %
Specific optical rotation +7.5° to +9.5° (20°C, 589 nm) (c=1 in CHCl3)
Residual solvents ≤10.0 % MTBE

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR