Imprimer

Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid 97%

Attention

ACRO434340010EA 236 CHF
ACRO434340010
Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid 97%
Pyrazolo[1,5-a]pyrimidine-2-carboxylic acid
Formule: C₇H₅N₃O₂
MW: 163,136 g/mol
Melting Pt: 289…291 °C
température de stockage: Température ambiante
MDL Number: MFCD00297221
Numéro CAS: 378211-85-9

Order Now

Résultats des tests de spécification

Appearance White to orange Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR