Imprimer

1,2-Dimethyl-1H-imidazole-5-carboxylic acid 97%

Attention

ACRO431160010EA 213 CHF
ACRO431160010 ACRO431160050
1,2-Dimethyl-1H-imidazole-5-carboxylic acid 97%
1,2-Dimethyl-1H-imidazole-5-carboxylic acid
Formule: C₆H₈N₂O₂
Melting Pt: 218,7…219.6 °C
température de stockage: Température ambiante
MDL Number: MFCD16659646
Numéro CAS: 122222-09-7

Order Now

Résultats des tests de spécification

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR