Imprimer

Aluminium oxide weakly acidic Brockmann Grade I 40-300 µm, 60A pour la chromatographie

ACRO392520050EA 400 CHF
ACRO392520050 ACRO392520010
Aluminium oxide weakly acidic Brockmann Grade I 40-300 µm, 60A pour la chromatographie
Aluminium oxide weakly acidic
Formule: Al₂O₃
MW: 101,96 g/mol
Point d'ébullition: 2980 °C (760 mmHg)
Melting Pt: 2050 °C
température de stockage: Température ambiante
MDL Number: MFCD00003424
Numéro CAS: 1344-28-1

Order Now

Résultats des tests de spécification

Appearance White to off-white Powder
Particle size ≤5 % (<40 µm)
Specific surface area 185 to 225 (m²/g)
pH 5.5 to 6.5 (10% aqueous suspension)
Sodium oxide (Na2O) ≤0.4 %
Silicon dioxide (SiO2) ≤0.03 %
Iron(III)oxide (Fe2O3) ≤0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR