Imprimer

1,1'-Ferrocenedimethanol 98%

Attention

ACRO382250050EA 501 CHF
ACRO382250050 ACRO382250010
1,1'-Ferrocenedimethanol 98%
1,1'-Ferrocenedimethanol
Formule: C₁₂H₁₄FeO₂
MW: 246,087 g/mol
Melting Pt: 106…109 °C
température de stockage: Température ambiante
MDL Number: MFCD02091688
Numéro CAS: 1291-48-1

Order Now

Résultats des tests de spécification

Appearance Yellow to orange to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 104 to 109 °C
Elemental Analysis ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR