3-Acetylphenanthrene 97%
|
Attention
|
ACRO102640050EA
110
CHF
ACRO102640050
ACRO102640250
3-Acetylphenanthrene 97%
3-Acetylphenanthrene
Formule:
C₁₆H₁₂O MW: 220,27 g/mol Melting Pt: 73…76 °C température de stockage: Température ambiante |
MDL Number:
MFCD00001169 Numéro CAS: 2039-76-1 |
Résultats des tests de spécification
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 71 to 76 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...