Amberjet® 4200-Cl
ACRO301335000EA
114
CHF
ACRO301335000
ACRO301330025
Amberjet® 4200-Cl
Amberjet® 4200-Cl
température de stockage:
Température ambiante |
MDL Number:
MFCD00145586 Numéro CAS: 60177-39-1 |
Résultats des tests de spécification
Appearance | Semitransparent yellow Beads |
Particle size |
≤0.5 % ( |
Particle size |
≤5.0 % ( |
Harmonic mean size | 0.600 to 0.800 mm |
Uniformity coefficient | ≤1.25 |
Moisture holding capacity | 49 to 55 % (Cl) |
Total volume capacity | ≥1.30 eq/l (Cl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...