Imprimer

5-Acetamido-2-(pentafluoroethyl)pyridine 96%

Attention

H66076.MCEA 183 CHF
H66076.MC
5-Acetamido-2-(pentafluoroethyl)pyridine 96%
5-Acetamido-2-(pentafluoroethyl)pyridine
Formule: C₉H₇F₅N₂O
MW: 254,16 g/mol
température de stockage: Température ambiante
MDL Number: MFCD28122877
Numéro CAS: 1580464-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR