Imprimer

4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester 96%

Fournisseur: Thermo Scientific

H62948.03EA 1190 CHF
H62948.03
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester 96%
4-(Boc-aminomethyl)-3-fluorobenzeneboronic acid pinacol ester
Formule: C₁₈H₂₇BFNO₄
MW: 351,22 g/mol
température de stockage: Température ambiante
MDL Number: MFCD22494714
Numéro CAS: 1351501-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR