3-Fluoro-2-methoxybenzeneboronic acid pinacol ester 96%
H62665.MDEA
359
CHF
H62665.MD
3-Fluoro-2-methoxybenzeneboronic acid pinacol ester 96%
3-Fluoro-2-methoxybenzeneboronic acid pinacol ester
Formule:
C₁₃H₁₈BFO₃ MW: 252,09 g/mol température de stockage: Température ambiante |
MDL Number:
MFCD16996297 Numéro CAS: 1417301-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...