Imprimer

3-Fluoro-4-(4-tetrahydropyranylamino)benzeneboronic acid pinacol ester ≥96%

H62624.MDEA 298 CHF
H62624.MD H62624.03
3-Fluoro-4-(4-tetrahydropyranylamino)benzeneboronic acid pinacol ester ≥96%
3-Fluoro-4-(4-tetrahydropyranylamino)benzeneboronic acid pinacol ester
Formule: C₁₇H₂₅BFNO₃
MW: 321,19 g/mol
température de stockage: Température ambiante
Numéro CAS: 950201-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR