Imprimer

3-Chloro-2-fluorobenzeneboronic acid pinacol ester ≥96%

H62521.MDEA 313 CHF
H62521.MD H62521.03
3-Chloro-2-fluorobenzeneboronic acid pinacol ester ≥96%
3-Chloro-2-fluorobenzeneboronic acid pinacol ester
Formule: C₁₂H₁₅BClFO₂
MW: 256,51 g/mol
température de stockage: Température ambiante
MDL Number: MFCD12405368
Numéro CAS: 1192025-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR