Imprimer

2-Carboxy-5-fluorobenzeneboronic acid pinacol ester ≥96%

H62390.MDEA 361 CHF
H62390.MD H62390.03
2-Carboxy-5-fluorobenzeneboronic acid pinacol ester ≥96%
2-Carboxy-5-fluorobenzeneboronic acid pinacol ester
Formule: C₁₃H₁₆BFO₄
MW: 266,07 g/mol
température de stockage: Température ambiante
MDL Number: MFCD18384083
Numéro CAS: 1191063-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR