Imprimer

3-Chloro-4-fluorobenzeneboronic acid pinacol ester ≥96%

H62307.03EA 923 CHF
H62307.03 H62307.MD
3-Chloro-4-fluorobenzeneboronic acid pinacol ester ≥96%
3-Chloro-4-fluorobenzeneboronic acid pinacol ester
Formule: C₁₂H₁₅BClFO₂
MW: 256,51 g/mol
température de stockage: Température ambiante
MDL Number: MFCD11111387
Numéro CAS: 635305-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR