Imprimer

3-Fluoro-5-formylbenzeneboronic acid pinacol ester ≥96%

H62303.03EA 923 CHF
H62303.03 H62303.MD
3-Fluoro-5-formylbenzeneboronic acid pinacol ester ≥96%
3-Fluoro-5-formylbenzeneboronic acid pinacol ester
Formule: C₁₃H₁₆BFO₃
MW: 250,07 g/mol
température de stockage: Température ambiante
MDL Number: MFCD16996301
Numéro CAS: 1112209-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR